Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006972.1 | NP_001006973.1 | 2 | 99 | ||
GenBank mRNA | BC083709.1 | - | 2 | 99 | 7190583 | |
FQ214688.1 | - | 2 | 106 | |||
FQ217076.1 | - | 2 | 157 |