Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007698.1 | NP_001007699.1 | 5 | 680 | ||
GenBank mRNA | BC081987.1 | - | 5 | 680 | 7129067 |