Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007803.1 | NP_001007804.1 | 10 | 1176 | ||
XM_006243250.1 | XP_006243312.1 | 9 | 1379 | |||
GenBank mRNA | BC085867.1 | - | 10 | 1302 | 7190926 |