Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008319.2 | NP_001008320.2 | 8 | 994 | ||
GenBank mRNA | BC085770.1 | - | 8 | 965 | 7189726 |