Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014699.3 | NP_055514.3 | 2 | 686 | ||
XM_005255710.3 | XP_005255767.1 | 2 | 1465 | |||
XM_005255711.3 | XP_005255768.1 | 2 | 1424 | |||
XM_005255712.3 | XP_005255769.1 | 2 | 1465 | |||
XM_011545990.2 | XP_011544292.1 | 2 | 691 | |||
GenBank mRNA | AB002294.1 | - | 1 | 820 | ||
AK304808.1 | - | 2 | 683 | |||
BC035589.1 | - | 2 | 672 | 5532753 |