Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270934.1 | NP_001257863.1 | 2, 3 | 678 | ||
NM_001270941.1 | NP_001257870.1 | 2, 3 | 678 | |||
NM_014790.4 | NP_055605.2 | 2, 3 | 678 | |||
XM_017010097.1 | XP_016865586.1 | 3, 4 | 672 | |||
XM_017010098.1 | XP_016865587.1 | 2, 3 | 678 | |||
GenBank mRNA | AB011127.1 | - | 2, 3 | 372 | ||
AF273057.1 | - | 2, 3 | 519 | |||
AK291280.1 | - | 2, 3 | 400 | |||
BC003189.1 | - | 2, 3 | 324 | |||
BC017354.2 | - | 2, 3 | 443 | 5020741 | ||
EF512550.1 | - | 1, 2 | 137 |