Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA0319 SIRNA Assays ›
Gene Symbol:
KIAA0319
Gene Name:
KIAA0319
Gene Aliases:
DYLX2, DYX2, NMIG
Chromosome Location:
Chr.6: 24540131 - 24646211 on Build GRCh38
UniGene ID:
Hs.26441
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22053
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA0319
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168374.1
NP_001161846.1
4
710
NM_001168375.1
NP_001161847.1
3
594
NM_001168376.1
NP_001161848.1
2
434
NM_001168377.1
NP_001161849.1
3
619
NM_014809.3
NP_055624.2
3
619
XM_011515025.2
XP_011513327.1
3
376
XM_011515026.2
XP_011513328.1
2
337
XM_017011541.1
XP_016867030.1
4
690
XM_017011542.1
XP_016867031.1
3
337
XM_017011543.1
XP_016867032.1
3
337
XM_017011544.1
XP_016867033.1
3
410
XM_017011545.1
XP_016867034.1
3
332
XM_017011546.1
XP_016867035.1
3
337
XM_017011547.1
XP_016867036.1
3
337
XM_017011548.1
XP_016867037.1
3
410
XM_017011549.1
XP_016867038.1
3
367
XM_017011550.1
XP_016867039.1
3
337
XM_017011551.1
XP_016867040.1
3
337
XM_017011552.1
XP_016867041.1
3
490
XM_017011553.1
XP_016867042.1
3
332
GenBank mRNA
AB002317.1
-
3
605
AK295008.1
-
2
171
AK296310.1
-
3
301
AK296426.1
-
4
445
AK307503.1
-
3
304
BC140821.1
-
4
450
9021689
BC144628.1
-
3
478
9053159
BC152460.1
-
3
605
8862596
DA316925.1
-
3
337
DA808234.1
-
2
420
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
negative regulation of dendrite development
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0