Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000965.1 | NP_001000965.1 | 1 | 619 |
Gene Symbol: LOC100912067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242832.2 | XP_006242894.1 | 1 | 619 |
Gene Symbol: LOC100912245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750483.3 | XP_003750531.1 | 619 |