Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001507.1 | NP_001001507.1 | 6, 7 | 1064 | ||
GenBank mRNA | AY353853.1 | - | 6, 7 | 1064 | ||
AY356356.1 | - | 6, 7 | 1064 |