Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014909.4 | NP_055724.1 | 7 | 2230 | ||
XM_017021087.1 | XP_016876576.1 | 9 | 1305 | |||
XM_017021088.1 | XP_016876577.1 | 8 | 1262 | |||
XM_017021089.1 | XP_016876578.1 | 8 | 1121 | |||
XM_017021090.1 | XP_016876579.1 | 7 | 1054 | |||
XM_017021091.1 | XP_016876580.1 | 7 | 2730 | |||
GenBank mRNA | AB028959.1 | - | 7 | 1685 | ||
AK291377.1 | - | 7 | 1380 | |||
AL832588.2 | - | 7 | 1934 |