Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213557.1 | NP_998722.1 | 3 | 276 | ||
GenBank mRNA | BC126072.1 | - | 3 | 250 | 8362450 | |
FQ209845.1 | - | 3 | 274 | |||
FQ210672.1 | - | 3 | 258 | |||
FQ210792.1 | - | 3 | 274 | |||
FQ217732.1 | - | 3 | 274 | |||
FQ220696.1 | - | 3 | 274 | |||
FQ221730.1 | - | 3 | 274 | |||
FQ221918.1 | - | 3 | 274 | |||
FQ221943.1 | - | 1 | 84 | |||
FQ222652.1 | - | 3 | 274 | |||
FQ222675.1 | - | 3 | 272 | |||
FQ223127.1 | - | 3 | 274 | |||
FQ223169.1 | - | 3 | 275 | |||
FQ223198.1 | - | 3 | 274 | |||
FQ223436.1 | - | 3 | 274 | |||
FQ223481.1 | - | 3 | 274 | |||
X57529.1 | - | 3 | 268 |
Gene Symbol: LOC100360679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727833.4 | XP_002727879.1 | 1 | 268 |