Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHA6 SIRNA Assays ›
Gene Symbol:
PLEKHA6
Gene Name:
pleckstrin homology domain containing A6
Gene Aliases:
PEPP-3, PEPP3
Chromosome Location:
Chr.1: 204218851 - 204378376 on Build GRCh38
UniGene ID:
Hs.253146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22432
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014935.4
NP_055750.2
8
1012
XM_005244966.4
XP_005245023.2
8
879
XM_005244967.4
XP_005245024.2
8
938
XM_005244968.4
XP_005245025.2
8
878
XM_006711212.3
XP_006711275.1
8
1165
XM_006711213.3
XP_006711276.2
8
939
XM_006711214.3
XP_006711277.2
8
939
XM_006711215.3
XP_006711278.2
8
939
XM_006711217.3
XP_006711280.2
8
939
XM_006711219.3
XP_006711282.2
8
939
XM_006711221.2
XP_006711284.1
8
1263
XM_006711222.3
XP_006711285.1
8
1248
XM_011509295.2
XP_011507597.1
8
939
XM_011509296.2
XP_011507598.1
8
939
XM_011509297.2
XP_011507599.1
10
1765
XM_011509298.2
XP_011507600.1
8
1048
XM_011509299.2
XP_011507601.2
8
1054
XM_011509301.2
XP_011507603.1
8
4944
XM_011509304.2
XP_011507606.1
8
893
XM_017000687.1
XP_016856176.1
8
939
XM_017000688.1
XP_016856177.1
8
939
XM_017000689.1
XP_016856178.1
8
879
XM_017000690.1
XP_016856179.1
8
4884
XM_017000691.1
XP_016856180.1
3
3337
XM_017000692.1
XP_016856181.1
3
3277
GenBank mRNA
AB023186.1
-
8
999
BC152475.1
-
8
999
8862612
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0