Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLEKHA6 SIRNA Assays ›
          Gene Symbol:
          PLEKHA6
          Gene Name:
          pleckstrin homology domain containing A6
          Gene Aliases:
          PEPP-3, PEPP3
          Chromosome Location:
          Chr.1: 204218851 - 204378376 on Build GRCh38
          UniGene ID:
          Hs.253146
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 22528

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLEKHA6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014935.4 NP_055750.2 16 2516
          XM_005244966.4 XP_005245023.2 20 2755
          XM_005244967.4 XP_005245024.2 16 2442
          XM_005244968.4 XP_005245025.2 16 2382
          XM_006711212.3 XP_006711275.1 20 3041
          XM_006711213.3 XP_006711276.2 19 2761
          XM_006711214.3 XP_006711277.2 20 2815
          XM_006711215.3 XP_006711278.2 18 2680
          XM_006711217.3 XP_006711280.2 19 2665
          XM_006711219.3 XP_006711282.2 17 2593
          XM_006711221.2 XP_006711284.1 20 3139
          XM_006711222.3 XP_006711285.1 20 3124
          XM_011509295.2 XP_011507597.1 20 2815
          XM_011509296.2 XP_011507598.1 19 2734
          XM_011509297.2 XP_011507599.1 22 3641
          XM_011509298.2 XP_011507600.1 20 2924
          XM_011509299.2 XP_011507601.2 20 2930
          XM_011509301.2 XP_011507603.1 20 6820
          XM_011509304.2 XP_011507606.1 20 2769
          XM_017000687.1 XP_016856176.1 20 2815
          XM_017000688.1 XP_016856177.1 19 2761
          XM_017000689.1 XP_016856178.1 18 2620
          XM_017000690.1 XP_016856179.1 20 6760
          XM_017000691.1 XP_016856180.1 15 5213
          XM_017000692.1 XP_016856181.1 15 5153
          GenBank mRNA AB023186.1 - 16 2503
          BC152475.1 - 16 2503 8862612
          EF491809.1 - 2 145

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0