Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159491.1 | NP_001152963.1 | 2 | 301 | ||
NM_015379.4 | NP_056194.1 | 2 | 301 | |||
XM_017011931.1 | XP_016867420.1 | 2 | 284 | |||
XM_017011932.1 | XP_016867421.1 | 2 | 284 | |||
XM_017011933.1 | XP_016867422.1 | 2 | 284 | |||
XM_017011934.1 | XP_016867423.1 | 2 | 284 | |||
XM_017011935.1 | XP_016867424.1 | 2 | 284 | |||
XM_017011936.1 | XP_016867425.1 | 2 | 295 | |||
GenBank mRNA | AB055977.1 | - | 2 | 293 | ||
AF041430.2 | - | 2 | 270 | |||
AF106966.1 | - | 2 | 252 | |||
BC018737.1 | - | 2 | 293 | 4875257 | ||
BC062370.1 | - | 2 | 265 | 6598528 | ||
BF108440.1 | - | 2 | 147 | |||
BF432881.1 | - | 2 | 146 |