Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003081.1 | 3 | 240 | |||
GenBank mRNA | AL050257.1 | - | 3 | 240 | ||
BC096726.1 | - | 3 | 240 | |||
BC098108.1 | - | 4 | 382 | |||
BC098364.1 | - | 3 | 240 | |||
BC099710.1 | - | 3 | 240 |
Gene Symbol: GSTTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127991.1 | - | 3 | 1396 |
Gene Symbol: LOC100652871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530666.1 | XP_011528968.1 | 3 | 299 |