Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H13 SIRNA Assays ›
Gene Symbol:
ZC3H13
Gene Name:
zinc finger CCCH-type containing 13
Gene Aliases:
KIAA0853
Chromosome Location:
Chr.13: 45954469 - 46052785 on Build GRCh38
UniGene ID:
Hs.136102
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241581
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015070.4
NP_055885.3
14
3830
XM_005266301.3
XP_005266358.1
15
4208
XM_005266302.3
XP_005266359.1
15
3908
XM_005266303.3
XP_005266360.1
16
4233
XM_005266304.3
XP_005266361.1
15
4190
XM_005266305.3
XP_005266362.1
16
4226
XM_005266306.3
XP_005266363.1
15
3904
XM_005266307.3
XP_005266364.1
15
4208
XM_005266308.3
XP_005266365.1
14
4124
XM_005266309.3
XP_005266366.1
14
3824
XM_005266310.3
XP_005266367.1
14
4106
XM_005266311.3
XP_005266368.1
14
4124
XM_005266312.1
XP_005266369.1
14
4124
XM_017020475.1
XP_016875964.1
16
3996
XM_017020476.1
XP_016875965.1
16
3993
XM_017020477.1
XP_016875966.1
15
4149
XM_017020478.1
XP_016875967.1
15
4142
XM_017020479.1
XP_016875968.1
15
3912
XM_017020480.1
XP_016875969.1
14
3820
XM_017020481.1
XP_016875970.1
15
4149
XM_017020482.1
XP_016875971.1
14
3824
GenBank mRNA
AB020660.1
-
3
1656
AK074924.1
-
1
884
AL136745.1
-
7
2866
AY283618.1
-
14
3828
CN262326.1
-
1
58
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026083, BC094575, BC138264, BC082576, BC023707.
Gene Ontology Categories:
Process(es)
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix