Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267597.1 | NP_001254526.1 | 6 | 471 | ||
NM_001267605.1 | NP_001254534.1 | 6 | 843 | |||
NM_001267606.1 | NP_001254535.1 | 4 | 1592 | |||
NM_001267607.1 | NP_001254536.1 | 6 | 843 | |||
NM_021045.2 | NP_066383.1 | 6 | 843 | |||
XM_006717485.3 | XP_006717548.1 | 5 | 800 | |||
XM_011519597.2 | XP_011517899.1 | 5 | 392 | |||
XM_011519598.2 | XP_011517900.1 | 6 | 493 | |||
XM_011519599.1 | XP_011517901.1 | 6 | 446 | |||
XM_011519600.2 | XP_011517902.1 | 5 | 334 | |||
XM_017016444.1 | XP_016871933.1 | 4 | 449 | |||
XM_017016445.1 | XP_016871934.1 | 5 | 940 | |||
XM_017016446.1 | XP_016871935.1 | 6 | 491 | |||
XM_017016447.1 | XP_016871936.1 | 4 | 449 | |||
XM_017016448.1 | XP_016871937.1 | 4 | 449 | |||
XM_017016449.1 | XP_016871938.1 | 5 | 789 | |||
XM_017016450.1 | XP_016871939.1 | 5 | 785 | |||
XM_017016451.1 | XP_016871940.1 | 5 | 940 | |||
GenBank mRNA | AJ491695.1 | - | 6 | 765 | ||
AJ492196.1 | - | 6 | 765 | |||
AK299357.1 | - | 6 | 471 | |||
BC021819.1 | - | 5 | 660 | |||
BC110869.1 | - | 4 | 1382 | |||
BC132915.1 | - | 6 | 501 | 40146937 | ||
BC132917.1 | - | 6 | 501 | 40146939 | ||
BC146820.1 | - | 5 | 340 | |||
DA128760.1 | - | 5 | 404 |