Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122842.2 | NP_001116314.1 | 16 | 5136 | ||
NM_001199619.1 | NP_001186548.1 | 17 | 5152 | |||
NM_001199620.1 | NP_001186549.1 | 18 | 5270 | |||
NM_001199621.1 | NP_001186550.1 | 15 | 4915 | |||
NM_001199622.1 | NP_001186551.1 | 6 | 2764 | |||
NM_181782.4 | NP_861447.3 | 16 | 5169 | |||
XM_005266822.4 | XP_005266879.1 | 16 | 5199 | |||
XM_006715340.3 | XP_006715403.1 | 16 | 5126 | |||
XM_011535455.2 | XP_011533757.1 | 16 | 5074 | |||
XM_017010269.1 | XP_016865758.1 | 16 | 4885 | |||
XM_017010270.1 | XP_016865759.1 | 17 | 5098 | |||
XM_017010271.1 | XP_016865760.1 | 16 | 5096 | |||
XM_017010272.1 | XP_016865761.1 | 16 | 4949 | |||
XM_017010273.1 | XP_016865762.1 | 16 | 5166 | |||
XM_017010274.1 | XP_016865763.1 | 16 | 5255 | |||
GenBank mRNA | AF493978.1 | - | 17 | 5151 | ||
AJ420542.1 | - | 8 | 3100 | |||
AK094706.1 | - | 6 | 2657 | |||
AL832621.2 | - | 16 | 4929 | |||
AL832628.2 | - | 16 | 4891 | |||
AL834442.1 | - | 14 | 4556 | |||
BC033612.1 | - | 9 | 3996 | |||
BX537385.1 | - | 16 | 4964 | |||
KC238672.1 | - | 6 | 2741 |