Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM49B SIRNA Assays ›
          Gene Symbol:
          FAM49B
          Gene Name:
          family with sequence similarity 49 member B
          Gene Aliases:
          BM-009, L1
          Chromosome Location:
          Chr.8: 129839593 - 130016697 on Build GRCh38
          UniGene ID:
          Hs.126941
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 24644

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM49B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256763.1 NP_001243692.1 6, 7 705
          NM_016623.4 NP_057707.3 5, 6 618
          NR_046359.1 8, 9 971
          NR_046360.1 8, 9 881
          NR_046361.1 7, 8 871
          XM_005250966.3 XP_005251023.1 4, 5 474
          XM_011517108.2 XP_011515410.1 7, 8 767
          XM_011517109.2 XP_011515411.1 7, 8 539
          XM_011517110.2 XP_011515412.1 6, 7 679
          XM_011517111.2 XP_011515413.1 6, 7 497
          XM_011517112.2 XP_011515414.1 8, 9 722
          XM_011517113.2 XP_011515415.1 7, 8 733
          XM_011517114.2 XP_011515416.1 7, 8 639
          XM_011517115.2 XP_011515417.1 6, 7 646
          XM_011517116.2 XP_011515418.1 7, 8 680
          XM_011517117.2 XP_011515419.1 6, 7 649
          XM_011517118.2 XP_011515420.1 6, 7 593
          XM_011517120.2 XP_011515422.1 5, 6 592
          XM_011517121.2 XP_011515423.1 8, 9 604
          XM_011517122.2 XP_011515424.1 6, 7 683
          XM_011517123.2 XP_011515425.1 6, 7 456
          XM_011517124.2 XP_011515426.1 5, 6 571
          XM_011517125.2 XP_011515427.1 5, 6 510
          XM_017013539.1 XP_016869028.1 8, 9 726
          XM_017013540.1 XP_016869029.1 8, 9 585
          XM_017013541.1 XP_016869030.1 6, 7 713
          XM_017013542.1 XP_016869031.1 8, 9 763
          XM_017013543.1 XP_016869032.1 8, 9 863
          XM_017013544.1 XP_016869033.1 8, 9 645
          XM_017013545.1 XP_016869034.1 9, 10 909
          XM_017013546.1 XP_016869035.1 7, 8 676
          XM_017013547.1 XP_016869036.1 9, 10 768
          XM_017013548.1 XP_016869037.1 7, 8 772
          XM_017013549.1 XP_016869038.1 7, 8 635
          XM_017013550.1 XP_016869039.1 6, 7 686
          XM_017013551.1 XP_016869040.1 8, 9 681
          XM_017013552.1 XP_016869041.1 8, 9 726
          XM_017013553.1 XP_016869042.1 7, 8 539
          XM_017013554.1 XP_016869043.1 9, 10 751
          XM_017013555.1 XP_016869044.1 6, 7 706
          XM_017013556.1 XP_016869045.1 6, 7 452
          XM_017013557.1 XP_016869046.1 7, 8 498
          XM_017013558.1 XP_016869047.1 7, 8 643
          XM_017013559.1 XP_016869048.1 7, 8 502
          XM_017013560.1 XP_016869049.1 5, 6 617
          XM_017013561.1 XP_016869050.1 5, 6 369
          XM_017013562.1 XP_016869051.1 6, 7 556
          GenBank mRNA AF208851.1 - 7, 8 558
          AK001226.1 - 1, 2 124
          AK002059.1 - 8, 9 681
          AK130372.1 - 5, 6 505
          AY364259.1 - 2, 3 173
          AY598320.1 - 3, 4 304
          BC003599.1 - 6, 7 484 3501828
          BC016345.1 - 7, 8 625 4041187
          BC017297.2 - 8, 9 853 5017493
          BX419336.2 - 6, 7 706
          CR749628.1 - 6, 7 451

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0