Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAIAP2 SIRNA Assays ›
Gene Symbol:
BAIAP2
Gene Name:
BAI1 associated protein 2
Gene Aliases:
BAP2, FLAF3, IRSP53
Chromosome Location:
Chr.17: 81035147 - 81117432 on Build GRCh38
UniGene ID:
Hs.128316
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25068
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAIAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144888.1
NP_001138360.1
10
1307
NM_006340.2
NP_006331.1
10
1307
NM_017450.2
NP_059344.1
10
1307
NM_017451.2
NP_059345.1
10
1307
XM_005256943.1
XP_005257000.1
11
1406
XM_005256944.1
XP_005257001.1
11
1406
XM_005256945.1
XP_005257002.1
11
1406
XM_005256948.3
XP_005257005.1
10
1310
XM_006721635.1
XP_006721698.1
11
1409
XM_006721636.1
XP_006721699.1
11
1409
XM_006721637.2
XP_006721700.1
10
1310
XM_011524193.1
XP_011522495.1
11
1409
XM_011524194.1
XP_011522496.1
11
1409
XM_011524195.1
XP_011522497.1
5
1069
XM_011524196.1
XP_011522498.1
5
831
XM_011524197.1
XP_011522499.1
5
828
XM_017024017.1
XP_016879506.1
9
2067
XM_017024018.1
XP_016879507.1
8
1529
XM_017024019.1
XP_016879508.1
5
1323
GenBank mRNA
AB015019.1
-
10
1292
AB015020.1
-
10
1292
AB017119.1
-
10
1292
AB017120.1
-
10
1292
AK023688.1
-
11
1392
AK056888.1
-
6
1479
AK222670.1
-
10
1303
AK225612.1
-
10
1304
AK297644.1
-
10
1236
AK301436.1
-
5
788
BC002495.2
-
10
1282
BC014020.2
-
10
1297
4125501
BC032559.1
-
10
1305
5562784
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
plasma membrane organization
axonogenesis
insulin receptor signaling pathway
regulation of cell shape
response to bacterium
dendrite development
positive regulation of actin filament polymerization
regulation of actin cytoskeleton organization
Fc-gamma receptor signaling pathway involved in phagocytosis
vascular endothelial growth factor receptor signaling pathway
regulation of synaptic plasticity
actin filament bundle assembly
actin crosslink formation
positive regulation of dendritic spine morphogenesis
cell-cell adhesion
positive regulation of actin cytoskeleton reorganization
protein binding
protein C-terminus binding
cytoskeletal adaptor activity
PDZ domain binding
identical protein binding
proline-rich region binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0