Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175268.4 | NP_780477.2 | 2 | 265 | ||
NM_212473.1 | NP_997638.1 | 2 | 328 | |||
GenBank mRNA | AK020830.1 | - | 2 | 266 | ||
AK051299.1 | - | 2 | 329 | |||
AK147350.1 | - | 2 | 661 | |||
AK147725.1 | - | 2 | 457 | |||
AK156629.1 | - | 2 | 228 | |||
AK165022.1 | - | 322 | ||||
AK178321.1 | - | 1 | 83 | |||
AK180944.1 | - | 1 | 82 | |||
AK187644.1 | - | 1 | 82 | |||
AK195672.1 | - | 1 | 33 | |||
AK207193.1 | - | 1 | 82 | |||
BC035523.1 | - | 2 | 260 | 5400742 |