Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009626.1 | NP_001009626.1 | 8 | 1108 | ||
GenBank mRNA | BC088180.1 | - | 8 | 1108 | 7364928 | |
FQ209491.1 | - | 8 | 1133 | |||
FQ209740.1 | - | 8 | 1144 | |||
FQ210182.1 | - | 8 | 1135 | |||
FQ210311.1 | - | 8 | 1134 | |||
FQ210756.1 | - | 8 | 1150 | |||
FQ211328.1 | - | 8 | 1107 | |||
FQ218068.1 | - | 3 | 421 | |||
FQ218335.1 | - | 4 | 641 | |||
FQ218445.1 | - | 8 | 1148 | |||
FQ218454.1 | - | 8 | 1135 | |||
FQ218597.1 | - | 8 | 1130 | |||
FQ218599.1 | - | 8 | 1134 | |||
FQ218637.1 | - | 8 | 1134 | |||
FQ218785.1 | - | 8 | 1151 | |||
FQ218859.1 | - | 8 | 1107 | |||
FQ218865.1 | - | 8 | 1134 | |||
FQ218961.1 | - | 8 | 1134 | |||
FQ219138.1 | - | 8 | 1199 | |||
FQ219328.1 | - | 8 | 1134 | |||
FQ219380.1 | - | 8 | 1135 | |||
FQ219438.1 | - | 8 | 1133 | |||
FQ219481.1 | - | 8 | 1146 | |||
FQ219537.1 | - | 8 | 1131 | |||
FQ219601.1 | - | 8 | 1147 | |||
X15551.1 | - | 8 | 963 |