Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BNC2 SIRNA Assays ›
Gene Symbol:
BNC2
Gene Name:
basonuclin 2
Gene Aliases:
BSN2
Chromosome Location:
Chr.9: 16409503 - 16870838 on Build GRCh38
UniGene ID:
Hs.656581
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25239
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BNC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001317939.1
NP_001304868.1
5
2492
NM_001317940.1
NP_001304869.1
5
2398
NM_017637.5
NP_060107.3
6
2618
XM_011517924.1
XP_011516226.1
5
2542
XM_011517934.2
XP_011516236.1
3
3570
XM_017014816.1
XP_016870305.1
8
4928
XM_017014817.1
XP_016870306.1
7
3077
XM_017014818.1
XP_016870307.1
6
2992
XM_017014819.1
XP_016870308.1
7
3076
XM_017014820.1
XP_016870309.1
7
3076
XM_017014821.1
XP_016870310.1
6
2990
XM_017014822.1
XP_016870311.1
4
2538
XM_017014823.1
XP_016870312.1
4
7307
XM_017014824.1
XP_016870313.1
3
2859
XM_017014825.1
XP_016870314.1
7
3075
XM_017014826.1
XP_016870315.1
7
3075
XM_017014827.1
XP_016870316.1
7
3075
XM_017014828.1
XP_016870317.1
7
3074
XM_017014829.1
XP_016870318.1
5
3046
GenBank mRNA
AK000050.1
-
1
402
AK025882.1
-
1
147
AK092247.1
-
1
1348
AK299073.1
-
5
2368
AK304745.1
-
3
2158
AY438376.1
-
4
2263
BX641030.1
-
1
1648
CR933649.1
-
5
2449
DQ884945.1
-
1
15
DQ884946.1
-
1
15
DQ884947.1
-
1
15
DQ884948.1
-
1
15
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endochondral bone growth
transcription, DNA-templated
regulation of transcription, DNA-templated
tongue development
palate development
mesenchyme development
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix