Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012190.1 | NP_001012190.1 | 4 | 1073 | ||
XM_008763626.1 | XP_008761848.1 | 4 | 1082 | |||
GenBank mRNA | BC083594.1 | - | 4 | 1073 | 7132783 |