Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC66 SIRNA Assays ›
Gene Symbol:
CCDC66
Gene Name:
coiled-coil domain containing 66
Gene Aliases:
-
Chromosome Location:
Chr.3: 56557125 - 56621837 on Build GRCh38
UniGene ID:
Hs.476399
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
255187
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC66
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012506.4
NP_001012524.4
14
2116
NM_001141947.1
NP_001135419.1
14
2135
NR_024460.1
14
2116
XM_005265081.3
XP_005265138.1
14
2153
XM_005265082.3
XP_005265139.1
14
2132
XM_005265083.3
XP_005265140.1
14
2024
XM_005265084.3
XP_005265141.1
14
2147
XM_011533614.2
XP_011531916.1
13
2158
XM_011533615.2
XP_011531917.1
13
2168
XM_011533616.2
XP_011531918.1
14
2092
XM_011533617.2
XP_011531919.1
15
2250
XM_011533619.2
XP_011531921.1
15
2153
XM_017006229.1
XP_016861718.1
14
2114
XM_017006230.1
XP_016861719.1
13
2136
XM_017006231.1
XP_016861720.1
13
2139
XM_017006232.1
XP_016861721.1
14
2006
XM_017006233.1
XP_016861722.1
14
1985
XM_017006234.1
XP_016861723.1
14
2174
XM_017006235.1
XP_016861724.1
15
2232
XM_017006236.1
XP_016861725.1
16
2245
XM_017006237.1
XP_016861726.1
16
2282
XM_017006238.1
XP_016861727.1
17
2423
XM_017006239.1
XP_016861728.1
11
1483
XM_017006240.1
XP_016861729.1
15
2186
XM_017006241.1
XP_016861730.1
16
2145
XM_017006242.1
XP_016861731.1
15
2008
XM_017006243.1
XP_016861732.1
14
2101
XM_017006244.1
XP_016861733.1
15
2237
XM_017006245.1
XP_016861734.1
13
1940
GenBank mRNA
AK096754.1
-
1
1451
AL832556.1
-
1
216
AL832692.1
-
14
2055
AL833326.1
-
15
2153
BC047509.1
-
14
2084
5276957
BC132827.1
-
14
2105
40146849
Back To Top
More Information
Gene Ontology Categories:
Function(s)
retinal rod cell development
detection of light stimulus involved in visual perception
post-embryonic retina morphogenesis in camera-type eye
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline