Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014150.3 | NP_001014172.3 | 3, 4 | 396 | ||
XM_006254664.2 | XP_006254726.1 | 12, 13 | 1557 | |||
GenBank mRNA | AY321347.1 | - | 12, 13 | 1563 | ||
BC083754.1 | - | 3, 4 | 396 |