Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb561a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014164.1 | NP_001014186.1 | 6 | 2272 | ||
XM_006231049.2 | XP_006231111.1 | 7 | 2371 | |||
GenBank mRNA | BC085835.1 | - | 6 | 2272 | 7134938 | |
FQ234846.1 | - | 1 | 562 |