Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024896.1 | NP_001020067.1 | 4 | 1816 | ||
XM_006255712.2 | XP_006255774.1 | 5 | 1800 | |||
XM_006255713.2 | XP_006255775.1 | 5 | 1593 | |||
XM_006255714.2 | XP_006255776.1 | 5 | 2753 | |||
XM_008772622.1 | XP_008770844.1 | 6 | 1815 | |||
GenBank mRNA | BC097457.1 | - | 4 | 1816 | 7457192 |