Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008863.3 | NP_001008863.2 | 8, 9 | 980 | ||
XM_008770560.1 | XP_008768782.1 | 11, 12 | 2343 |