Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147153.3 | NP_671495.1 | 18 | 1907 | ||
NM_178851.3 | NP_849182.1 | 17 | 1874 | |||
GenBank mRNA | AB028844.1 | - | 17 | 1854 | ||
AF281050.1 | - | 18 | 1884 | |||
AF281051.1 | - | 17 | 1851 | |||
AK038824.1 | - | 17 | 1863 | |||
AK042253.1 | - | 17 | 1863 | |||
AK084743.1 | - | 17 | 1874 | |||
AK129210.1 | - | 17 | 1869 | |||
AK140041.1 | - | 18 | 1912 | |||
AK141424.1 | - | 17 | 1857 | |||
AK159889.1 | - | 17 | 1867 | |||
AK160109.1 | - | 17 | 1867 | |||
BC007479.1 | - | 17 | 1825 | 3492875 |