Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012079.1 | NP_001012079.1 | 22 | 3221 | ||
XM_006232643.2 | XP_006232705.1 | 27 | 3999 | |||
XM_006232644.2 | XP_006232706.1 | 28 | 4116 | |||
XM_006232646.2 | XP_006232708.1 | 26 | 3576 | |||
XM_006232647.2 | XP_006232709.1 | 25 | 3420 | |||
XM_006232648.2 | XP_006232710.1 | 25 | 3371 | |||
XM_006232651.2 | XP_006232713.1 | 22 | 3509 | |||
XM_006232652.2 | XP_006232714.1 | 22 | 3328 | |||
XM_006232653.2 | XP_006232715.1 | 22 | 3216 | |||
XM_006232654.2 | XP_006232716.1 | 22 | 4025 | |||
XM_008761151.1 | XP_008759373.1 | 28 | 4111 | |||
XM_008761152.1 | XP_008759374.1 | 23 | 4099 | |||
GenBank mRNA | BC090078.1 | - | 22 | 3221 | 7125049 | |
FQ212011.1 | - | 23 | 3194 |