Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhobtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013133.1 | NP_001013151.1 | 4 | 738 | ||
GenBank mRNA | BC091309.1 | - | 4 | 738 | 7311705 |