Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015124.4 | NP_055939.1 | 14 | 1384 | ||
XM_005261396.3 | XP_005261453.1 | 14 | 1320 | |||
XM_006724169.1 | XP_006724232.1 | 14 | 1282 | |||
XM_006724170.3 | XP_006724233.1 | 14 | 1301 | |||
XM_011530011.1 | XP_011528313.1 | 13 | 1220 | |||
XM_011530012.1 | XP_011528314.1 | 13 | 1220 | |||
XM_011530013.2 | XP_011528315.2 | 14 | 1329 | |||
XM_011530014.2 | XP_011528316.1 | 13 | 1220 | |||
XM_017028670.1 | XP_016884159.1 | 14 | 1342 | |||
GenBank mRNA | AB018310.2 | - | 14 | 1384 | ||
BC129837.1 | - | 13 | 1205 | 40119070 | ||
CR456470.1 | - | 13 | 1210 |