Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025136.1 | NP_001020307.1 | 1 | 1275 | ||
GenBank mRNA | BC087133.1 | - | 1 | 1275 | 7122255 |