Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178457.2 | NP_848552.1 | 5 | 5817 | ||
XM_005260272.3 | XP_005260329.1 | 6 | 6756 | |||
XM_005260273.3 | XP_005260330.1 | 7 | 8830 | |||
XM_006723698.3 | XP_006723761.1 | 6 | 6153 | |||
XM_011528534.2 | XP_011526836.1 | 7 | 7397 | |||
XM_011528536.2 | XP_011526838.1 | 6 | 6579 | |||
XM_011528537.2 | XP_011526839.1 | 7 | 6840 | |||
XM_011528538.2 | XP_011526840.1 | 6 | 6922 | |||
XM_017027642.1 | XP_016883131.1 | 6 | 7211 | |||
XM_017027643.1 | XP_016883132.1 | 7 | 6237 | |||
XM_017027644.1 | XP_016883133.1 | 7 | 6275 | |||
GenBank mRNA | AL713683.1 | - | 1 | 1401 | ||
AL833682.1 | - | 5 | 4686 |