Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178457.2 | NP_848552.1 | 5 | 5085 | ||
XM_005260272.3 | XP_005260329.1 | 6 | 6024 | |||
XM_005260273.3 | XP_005260330.1 | 7 | 8098 | |||
XM_006723698.3 | XP_006723761.1 | 6 | 5421 | |||
XM_011528534.2 | XP_011526836.1 | 7 | 6665 | |||
XM_011528536.2 | XP_011526838.1 | 6 | 5847 | |||
XM_011528537.2 | XP_011526839.1 | 7 | 6108 | |||
XM_011528538.2 | XP_011526840.1 | 6 | 6190 | |||
XM_017027642.1 | XP_016883131.1 | 6 | 6479 | |||
XM_017027643.1 | XP_016883132.1 | 7 | 5505 | |||
XM_017027644.1 | XP_016883133.1 | 7 | 5543 | |||
GenBank mRNA | AL713683.1 | - | 1 | 669 | ||
AL833682.1 | - | 5 | 3954 |