Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DZANK1 SIRNA Assays ›
          Gene Symbol:
          DZANK1
          Gene Name:
          double zinc ribbon and ankyrin repeat domains 1
          Gene Aliases:
          ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
          Chromosome Location:
          Chr.20: 18383367 - 18467185 on Build GRCh38
          UniGene ID:
          Hs.472225
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 26401

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DZANK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099407.1 NP_001092877.1 14, 15 1743
          XM_005260742.3 XP_005260799.1 14, 15 1795
          XM_005260743.3 XP_005260800.1 13, 14 1649
          XM_006723584.2 XP_006723647.1 12, 13 1570
          XM_011529268.2 XP_011527570.1 14, 15 1913
          XM_011529271.2 XP_011527573.1 14, 15 2526
          XM_011529272.2 XP_011527574.1 14, 15 1909
          XM_011529275.2 XP_011527577.1 13, 14 2399
          XM_011529277.2 XP_011527579.1 6, 7 2640
          XM_017027909.1 XP_016883398.1 14, 15 1896
          XM_017027910.1 XP_016883399.1 14, 15 1779
          XM_017027911.1 XP_016883400.1 14, 15 1775
          XM_017027912.1 XP_016883401.1 14, 15 2482
          XM_017027913.1 XP_016883402.1 14, 15 2467
          XM_017027914.1 XP_016883403.1 14, 15 1721
          XM_017027915.1 XP_016883404.1 14, 15 1717
          XM_017027916.1 XP_016883405.1 12, 13 2281
          XM_017027917.1 XP_016883406.1 15, 16 2576
          XM_017027918.1 XP_016883407.1 15, 16 1830
          XM_017027919.1 XP_016883408.1 15, 16 1826
          XM_017027920.1 XP_016883409.1 13, 14 1785
          XM_017027921.1 XP_016883410.1 7, 8 947
          GenBank mRNA AK001462.1 - 8, 9 930
          AK055544.1 - 13, 14 1538
          AK131451.1 - 15, 16 2382
          AK295073.1 - 14, 15 1653
          AK299746.1 - 14, 15 1666
          BC130341.1 - 13, 14 1571 40146372
          BC144153.1 - 13, 14 1571 9052673
          CR749327.1 - 15, 16 1795
          DQ104738.1 - 5, 6 664

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy in response to mitochondrial depolarization
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix