Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UEVLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040697.2 | NP_001035787.1 | 4 | 419 | ||
NM_001261382.1 | NP_001248311.1 | 3 | 353 | |||
NM_001261383.1 | NP_001248312.1 | 3 | 353 | |||
NM_001261385.1 | NP_001248314.1 | 4 | 376 | |||
NM_001297771.1 | NP_001284700.1 | 4 | 419 | |||
NM_018314.4 | NP_060784.3 | 4 | 419 | |||
XM_006718255.2 | XP_006718318.1 | 3 | 285 | |||
GenBank mRNA | AF503350.1 | - | 4 | 379 | ||
AK001930.1 | - | 4 | 376 | |||
AK296835.1 | - | 3 | 313 | |||
AK301537.1 | - | 3 | 322 | |||
AK314521.1 | - | 4 | 419 | |||
BC011011.1 | - | 4 | 356 | |||
BC064566.1 | - | 3 | 293 | 5576632 | ||
CB991602.1 | - | 4 | 434 |