Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM153A SIRNA Assays ›
          Gene Symbol:
          FAM153A
          Gene Name:
          family with sequence similarity 153 member A
          Gene Aliases:
          NY-REN-7
          Chromosome Location:
          Chr.5: 177694247 - 177783518 on Build GRCh38
          UniGene ID:
          Hs.368516
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 267251

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM153A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005265889.3 XP_005265946.1 28 2755
          XM_006714848.3 XP_006714911.1 29 3336
          XM_006714849.3 XP_006714912.1 28 3161
          XM_011534520.2 XP_011532822.1 29 3358
          XM_011534521.2 XP_011532823.1 30 3401
          XM_011534522.2 XP_011532824.1 30 3485
          XM_011534523.2 XP_011532825.1 29 3426
          XM_011534528.2 XP_011532830.1 21 2968
          XM_017009352.1 XP_016864841.1 29 3364
          XM_017009353.1 XP_016864842.1 29 2942
          XM_017009354.1 XP_016864843.1 29 2902
          XM_017009355.1 XP_016864844.1 29 3369
          XM_017009356.1 XP_016864845.1 29 2872
          XM_017009357.1 XP_016864846.1 29 2927
          XM_017009360.1 XP_016864849.1 29 2836
          XM_017009361.1 XP_016864850.1 28 2818
          XM_017009362.1 XP_016864851.1 28 3208
          XM_017009363.1 XP_016864852.1 30 3285
          XM_017009364.1 XP_016864853.1 30 3442
          XM_017009365.1 XP_016864854.1 30 3279
          XM_017009366.1 XP_016864855.1 30 3454
          XM_017009367.1 XP_016864856.1 29 3332
          XM_017009368.1 XP_016864857.1 29 3410
          XM_017009369.1 XP_016864858.1 29 3349
          XM_017009370.1 XP_016864859.1 29 3334
          XM_017009371.1 XP_016864860.1 28 3162
          GenBank mRNA AB018295.1 - 27 3013

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0