Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023073.3 | NP_075561.3 | 11 | 1721 | ||
XM_005248345.3 | XP_005248402.1 | 11 | 1709 | |||
XM_005248346.3 | XP_005248403.1 | 11 | 1629 | |||
XM_005248347.3 | XP_005248404.1 | 11 | 1629 | |||
XM_005248349.3 | XP_005248406.1 | 11 | 1629 | |||
XM_005248350.3 | XP_005248407.1 | 11 | 1627 | |||
XM_011514085.2 | XP_011512387.1 | 11 | 1628 | |||
XM_011514086.2 | XP_011512388.1 | 11 | 1625 | |||
XM_011514087.2 | XP_011512389.1 | 11 | 1714 | |||
XM_011514088.2 | XP_011512390.1 | 11 | 1714 | |||
XM_011514089.2 | XP_011512391.1 | 11 | 1714 | |||
XM_011514090.2 | XP_011512392.1 | 11 | 1492 | |||
XM_011514092.2 | XP_011512394.1 | 11 | 1714 | |||
XM_017009760.1 | XP_016865249.1 | 10 | 1440 | |||
XM_017009761.1 | XP_016865250.1 | 10 | 1408 | |||
XM_017009762.1 | XP_016865251.1 | 7 | 1167 | |||
XM_017009763.1 | XP_016865252.1 | 7 | 1040 | |||
XM_017009764.1 | XP_016865253.1 | 7 | 1001 | |||
XM_017009765.1 | XP_016865254.1 | 11 | 1409 | |||
GenBank mRNA | DN831769.1 | - | 1 | 67 |