Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120426.1 | 5 | 613 | |||
GenBank mRNA | HY006332.1 | - | 4 | 313 |
Gene Symbol: LOC102723968 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130763.1 | 5 | 1206 | |||
GenBank mRNA | AK098560.1 | - | 5 | 1206 | ||
AK127969.1 | - | 4 | 2220 |