Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM153A SIRNA Assays ›
          Gene Symbol:
          FAM153A LOC100507387
          Gene Name:
          family with sequence similarity 153 member A
          uncharacterized LOC100507387
          Gene Aliases:
          NY-REN-7
          -
          Chromosome Location:
          Chr.5: 177694247 - 177783518 on Build GRCh38
          Chromosome Location:
          Chr.5: 176119549 - 176125165 on Build GRCh38
          UniGene ID:
          Hs.368516
          Hs.368516
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 275115

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM153A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005265889.3 XP_005265946.1 23 1564
          XM_006714848.3 XP_006714911.1 23 1958
          XM_006714849.3 XP_006714912.1 22 1783
          XM_011534520.2 XP_011532822.1 23 1980
          XM_011534521.2 XP_011532823.1 24 2023
          XM_011534522.2 XP_011532824.1 24 2107
          XM_011534523.2 XP_011532825.1 23 2048
          XM_011534528.2 XP_011532830.1 15 1590
          XM_017009352.1 XP_016864841.1 23 1986
          XM_017009353.1 XP_016864842.1 23 1564
          XM_017009354.1 XP_016864843.1 23 1524
          XM_017009355.1 XP_016864844.1 23 1986
          XM_017009357.1 XP_016864846.1 23 1564
          XM_017009358.1 XP_016864847.1 23 1564
          XM_017009359.1 XP_016864848.1 23 1564
          XM_017009360.1 XP_016864849.1 23 1564
          XM_017009361.1 XP_016864850.1 23 1564
          XM_017009362.1 XP_016864851.1 22 1830
          XM_017009363.1 XP_016864852.1 24 1907
          XM_017009364.1 XP_016864853.1 24 2064
          XM_017009365.1 XP_016864854.1 24 1901
          XM_017009366.1 XP_016864855.1 24 2076
          XM_017009367.1 XP_016864856.1 23 1954
          XM_017009368.1 XP_016864857.1 23 2032
          XM_017009369.1 XP_016864858.1 23 1971
          XM_017009370.1 XP_016864859.1 23 1971
          XM_017009371.1 XP_016864860.1 23 1971
          Gene Symbol: LOC100507387
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_038402.1 1 74
          GenBank mRNA AL117630.1 - 1 72
          DA164303.1 - 1 133

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0