Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF346 SIRNA Assays ›
Gene Symbol:
ZNF346
Gene Name:
zinc finger protein 346
Gene Aliases:
JAZ, Zfp346
Chromosome Location:
Chr.5: 177022680 - 177081189 on Build GRCh38
UniGene ID:
Hs.484259
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
2780
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF346
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308213.1
NP_001295142.1
8
987
NM_001308214.1
NP_001295143.1
6
816
NM_001308215.1
NP_001295144.1
6
726
NM_001308216.1
NP_001295145.1
4
487
NM_001308218.1
NP_001295147.1
4
618
NM_001308219.1
NP_001295148.1
4
487
NM_001308221.1
NP_001295150.1
3
384
NM_001308223.1
NP_001295152.1
3
384
NM_012279.3
NP_036411.1
7
912
NR_131773.1
6
767
NR_131774.1
5
581
XM_005265860.2
XP_005265917.1
8
980
XM_005265864.3
XP_005265921.1
7
746
XM_011534493.1
XP_011532795.1
8
980
XM_011534494.1
XP_011532796.1
7
886
XM_011534495.2
XP_011532797.1
7
886
XM_011534496.2
XP_011532798.1
7
838
XM_017009294.1
XP_016864783.1
7
960
XM_017009295.1
XP_016864784.1
7
912
XM_017009296.1
XP_016864785.1
7
912
XM_017009297.1
XP_016864786.1
6
866
XM_017009299.1
XP_016864788.1
6
818
XM_017009300.1
XP_016864789.1
7
1250
XM_017009301.1
XP_016864790.1
6
726
XM_017009302.1
XP_016864791.1
8
925
XM_017009303.1
XP_016864792.1
5
1023
GenBank mRNA
AF083340.1
-
7
885
AK026627.1
-
7
880
AK131469.1
-
8
932
AK295549.1
-
6
792
AK297448.1
-
4
602
AK297584.1
-
3
368
AK300065.1
-
6
731
AK300738.1
-
5
526
AL359576.1
-
7
860
BC007775.2
-
7
867
4301820
BU860103.1
-
6
701
BX331331.2
-
4
442
CR749692.1
-
7
860
DA169130.1
-
4
460
DR003225.1
-
3
387
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of apoptotic process
intrinsic apoptotic signaling pathway by p53 class mediator
p53 binding
double-stranded RNA binding
protein binding
zinc ion binding
enzyme binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline