Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP170B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112726.2 | NP_001106197.1 | 19 | 5769 | ||
NM_015005.2 | NP_055820.2 | 18 | 5742 | |||
XM_005267550.3 | XP_005267607.1 | 19 | 5856 | |||
XM_011536665.2 | XP_011534967.1 | 20 | 6009 | |||
XM_011536666.2 | XP_011534968.1 | 19 | 6372 | |||
XM_011536667.2 | XP_011534969.1 | 12 | 4048 | |||
XM_017021227.1 | XP_016876716.1 | 18 | 6621 | |||
XM_017021228.1 | XP_016876717.1 | 18 | 6516 | |||
GenBank mRNA | AB006622.3 | - | 19 | 5769 | ||
AK025023.1 | - | 4 | 1344 | |||
BC047913.1 | - | 5 | 1437 | |||
BC112928.1 | - | 18 | 5773 | 6141521 |