Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020205.3 | NP_064590.2 | 2, 3 | 449 | ||
XM_011509782.1 | XP_011508084.1 | 2, 3 | 446 | |||
XM_011509783.2 | XP_011508085.1 | 2, 3 | 676 | |||
XM_011509784.2 | XP_011508086.1 | 2, 3 | 240 | |||
XM_011509785.2 | XP_011508087.1 | 2, 3 | 3873 | |||
XM_011509786.1 | XP_011508088.1 | 2, 3 | 446 | |||
XM_011509787.1 | XP_011508089.1 | 2, 3 | 189 | |||
XM_017001850.1 | XP_016857339.1 | 2, 3 | 446 | |||
GenBank mRNA | AJ293573.1 | - | 2, 3 | 292 | ||
AK299790.1 | - | 2, 3 | 435 | |||
BC020622.1 | - | 2, 3 | 235 | |||
BC072681.1 | - | 2, 3 | 386 | 6170513 | ||
BP321045.1 | - | 2, 3 | 449 | |||
BP357326.1 | - | 2, 3 | 449 | |||
BX641079.1 | - | 1 | 18 |