Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030044.1 | NP_001025215.1 | 3 | 1406 | ||
XM_006255197.2 | XP_006255259.1 | 3 | 1189 | |||
XM_008772318.1 | XP_008770540.1 | 3 | 1178 | |||
GenBank mRNA | DQ109812.1 | - | 3 | 1406 |