Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLSCR2 SIRNA Assays ›
Gene Symbol:
PLSCR2
Gene Name:
phospholipid scramblase 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 146391363 - 146497163 on Build GRCh38
UniGene ID:
Hs.744414
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28368
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLSCR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199978.1
NP_001186907.1
8
1185
NM_001199979.1
NP_001186908.1
6
802
NM_020359.2
NP_065092.1
7
915
XM_011513013.2
XP_011511315.2
6
1965
XM_011513019.2
XP_011511321.2
6
1965
XM_011513020.2
XP_011511322.2
6
1965
XM_011513021.2
XP_011511323.2
6
1965
XM_011513022.2
XP_011511324.2
6
1965
XM_011513023.2
XP_011511325.2
6
1965
XM_017006898.1
XP_016862387.1
6
1965
XM_017006899.1
XP_016862388.1
6
1965
XM_017006900.1
XP_016862389.1
6
1965
XM_017006901.1
XP_016862390.1
6
1965
XM_017006902.1
XP_016862391.1
6
1965
XM_017006903.1
XP_016862392.1
6
1965
XM_017006904.1
XP_016862393.1
6
1965
XM_017006905.1
XP_016862394.1
8
1421
XM_017006906.1
XP_016862395.1
8
1202
XM_017006907.1
XP_016862396.1
8
967
XM_017006908.1
XP_016862397.1
8
1092
XM_017006909.1
XP_016862398.1
7
879
XM_017006910.1
XP_016862399.1
6
992
XM_017006911.1
XP_016862400.1
7
1269
XM_017006912.1
XP_016862401.1
6
620
XM_017006913.1
XP_016862402.1
4
1591
XM_017006914.1
XP_016862403.1
6
827
XM_017006915.1
XP_016862404.1
6
809
GenBank mRNA
AF159441.1
-
7
859
AK301909.1
-
8
1185
BC055415.1
-
6
806
BC069785.1
-
6
752
7262547
BC120969.2
-
6
824
40115378
BC141969.1
-
6
824
40115468
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
phospholipid scrambling
calcium ion binding
phospholipid scramblase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0