Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017381.2 | NP_059077.1 | 10 | 1488 | ||
GenBank mRNA | AK146275.1 | - | 10 | 1486 | ||
AK150291.1 | - | 10 | 1527 | |||
AK152536.1 | - | 10 | 1527 | |||
AK160862.1 | - | 9 | 1376 | |||
AK166534.1 | - | 10 | 1483 | |||
AK167756.1 | - | 10 | 1485 | |||
AK167766.1 | - | 10 | 1516 | |||
AK167862.1 | - | 10 | 1516 | |||
AK167983.1 | - | 10 | 1516 | |||
AK168033.1 | - | 10 | 1485 | |||
AK168162.1 | - | 10 | 1487 | |||
AK169114.1 | - | 10 | 1480 | |||
AK169335.1 | - | 10 | 1489 | |||
BC049115.1 | - | 10 | 1498 | |||
BC051187.1 | - | 5 | 1075 | 6478334 | ||
BC132547.1 | - | 10 | 1485 | 40130824 | ||
BC138575.1 | - | 10 | 1485 | 8861595 |