Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014099.1 | NP_001014121.1 | 3 | 934 | ||
GenBank mRNA | BC079247.1 | - | 3 | 934 | 7132885 | |
DQ088964.1 | - | 1 | 481 |