Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033961.1 | NP_001029133.1 | 8 | 1532 | ||
NM_020094.2 | NP_064479.2 | 10 | 1682 | |||
NM_020095.2 | NP_064480.2 | 9 | 1628 | |||
XM_008760448.1 | XP_008758670.1 | 8 | 3240 | |||
XM_008760449.1 | XP_008758671.1 | 10 | 1791 | |||
XM_008760450.1 | XP_008758672.1 | 8 | 1432 | |||
GenBank mRNA | BC085905.1 | - | 8 | 1532 | 7193750 |
Gene Symbol: LOC100911951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231581.2 | XP_006231643.1 | 8 | 1428 | ||
XM_008760479.1 | XP_008758701.1 | 10 | 1739 | |||
XM_008760480.1 | XP_008758702.1 | 9 | 1684 | |||
XM_008760481.1 | XP_008758703.1 | 8 | 1746 | |||
XM_008760482.1 | XP_008758704.1 | 8 | 1587 | |||
XM_008760483.1 | XP_008758705.1 | 10 | 1791 |